Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6690 Cooling Arm Sleeves

Catalog No. 19819637 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-637 2X-Large
19-819-634 Medium
19-819-635 Large
19-819-636 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-819-637 Supplier Ergodyne™ Supplier No. 12386
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chill-Its™ 6690 Cooling Arm Sleeves provide full arm coverage to absorb and wick away sweat, deliver advanced cooling relief, shield skin from UV rays, and even protect against light abrasions.

  • Performance knit fabric is soft, stretchable and durable
  • Instant cooling when wet that can last for hours
  • Wicks moisture when dry to absorb sweat from the body
  • UPF 50+ Protection
  • Protection from light abrasions
  • Reflective accents
  • Anti-odor treatment
  • Machine washable
  • Chemical-free cooling

Specifications

Material Performance Knit Fabric
Product Type Arm Sleeves
Size 2X-Large
Color Black
For Use With (Application) Agriculture, Construction, Utilities, Transportation, Sports
Quantity 1/Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.