Learn More
Ergodyne™ Chill-Its™ 6700FR Evaporative FR Cooling Tie Bandana
Catalog No. 19819586 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
Lime
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-586 Blue
19-819-584 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-819-586 Supplier Ergodyne™ Supplier No. 12607
FR Evaporative Cooling Bandana featured activated polymers that provide cooling relief and FR protection.
  • Modacrylic cotton blend fabric
  • Fabric meets ASTM F1506 performance requirements (ATPV rating of 7.1 cal/cm2); classified CAT1 per NFPA 70E.
  • Meets ASTM F2302 for flame resistance
  • Soak in water 2-5 minutes to activate
  • Remains hydrated for up to 4 hours
Specifications
Material Modacrylic Cotton Blend
Product Type Bandana
Color Blue