Learn More
Description
- Soak in water 2-5 minutes to activate
- Remains hydrated for up to 4 hours
- Reusable
- Available in a variety of colors and patterns like red western, blue western, camo, stars and striped, black, lime, blue, skulls, and flames
Specifications
Specifications
| Color | Black |
| Closure Type | Hook and Loop |
| For Use With (Application) | Construction, Maintenance, Landscaping, Material Handling |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.