Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6705 Evaporative Cooling Bandana with Hook and Loop

Catalog No. 19819506 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Blue, White
Camouflage
Gray, Black, White
Lime
Red, White
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-506 Gray, Black, White
19-819-483 Camouflage
19-819-484 Red, White
19-819-485 Blue, White
19-819-486 Lime
19-819-497 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-819-506 Supplier Ergodyne™ Supplier No. 12349
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Evaporative Cooling Bandana features activated polymers that bring cooling power to a variety of application.

  • Soak in water 2-5 minutes to activate
  • Remains hydrated for up to 4 hours
  • Reusable
  • Available in a variety of colors and patterns like red western, blue western, camo, stars and striped, black, lime, blue, skulls, and flames

Specifications

Product Type Bandana
Color Gray, Black, White
Closure Type Hook and Loop
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.