Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6705CT Evaporative Hook and Loop Bandana with Cooling Towel

Catalog No. 19819559 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Blue
Blue, White
Lime
Red, White, Blue
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-559 Blue, White
19-819-560 Lime
19-819-556 Red, White, Blue
19-819-561 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-819-559 Supplier Ergodyne™ Supplier No. 12574
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chill-Its™ 6705CT Evaporative Hook and Loop Bandana with Cooling Towel features advanced PVA material that keeps you cool without the bulk.

  • Advanced PVA material activates easily and quickly when run under water for one minute
  • Reusable
  • Super-evaporative cooling process helps keep you cool
  • Breakaway hook and loop closure
  • Available in a variety of colors and patterns like red western, blue western, camo, stars and striped, black, lime, blue, skulls, and flames

Specifications

Material PVA
Product Type Bandana
Color Blue, White
Closure Type Hook and Loop
For Use With (Application) Construction, Maintenance, Construction, Landscaping, Material Handling
Includes Cooling Towel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.