Learn More
Ergodyne™ Chill-Its™ 6705FR Evaporative FR Cooling Bandana with Hook and Loop
Shop All Ergodyne ProductsDescription
- Modacrylic cotton blend fabric
- Fabric meets ASTM F1506 performance requirements (ATPV rating of 7.1 cal/cm2); classified CAT1 per NFPA 70E.
- Meets ASTM F2302 for flame resistance
- Soak in water 2-5 minutes to activate
- Remains hydrated for up to 4 hours
- Reusable
Specifications
Specifications
| Material | Modacrylic Cotton Blend |
| Product Type | Bandana |
| Color | Blue |
| Closure Type | Hook and Loop |
| Certifications/Compliance | ASTM F1506, ASTM F2302 |
| For Use With (Application) | Landscaping, Material Handling, Utility Crews, Electrical Workers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.