Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6715 Evaporative Cooling Hard Hat Pad

Catalog No. 19819502 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-819-502 Color: Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-502 Supplier Ergodyne™ Supplier No. 12337
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chill-Its™ 6715 Evaporative Cooling Hard Hat Pad attaches to standard hard hat headbands.

  • Lightweight, cooling acrylic polymers
  • Hook and loop closures
  • Conveniently attaches to hard hat suspension headband
  • Soak in water 2 to 5 minutes to activate
  • Remains hydrated for up to 4 hours
  • Reactivate by re-soaking

Specifications

Color Blue
For Use With (Application) Construction, Maintenance, Landscaping, Warehousing/Distribution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.