Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 6715CT Evaporative Cooling Hard Hat Pad

Catalog No. 19819652 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-819-652 Color: Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-819-652 Supplier Ergodyne™ Supplier No. 12597
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chill-Its™ 6715CT Evaporative Cooling Hard Hat Pad attaches to standard hard hat headbands.

  • Advanced PVA material activates easily and quickly when run under water for one minute
  • Hook and loop closures
  • Conveniently attaches to hard hat suspension headband
  • Reusable

Specifications

Color Blue
For Use With (Application) Construction, Landscaping
Material PVA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.