Learn More
Ergodyne™ Chill-Its™ 8935CT Evaporative Class Headwear Hi-Vis Ranger Hat with Cooling Towel
Catalog No. 19819574 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-574 Small/Medium
19-819-575 Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-819-574 Supplier Ergodyne™ Supplier No. 12590
Add to Cart
Add to Cart
Ranger hat with wide brim and vented mesh side panels is lined with an advanced PVA material for cooling relief.
  • Advanced PVA material lines the inside hat rim for relief from the heat
  • Easily activated by running under water for one minute
  • Cooling sensation lasts for up to 4 hours
  • ANSI certified 150D Oxford Polyester
  • ANSI-compliant tape with contrasting color
Specifications
Material PVA-lined Polyester
Product Type Ranger Hat
Size Small/Medium
Color Lime