Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Chill-Its™ 8935MF Evaporative Class Headwear Hi-Vis Ranger Hat with Microfiber Lining

Catalog No. 19819583 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-583 -
19-819-576 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-819-583 Supplier Ergodyne™ Supplier No. 12595
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ranger hat with wide brim and vented mesh side panels is lined with an advanced microfiber material for cooling relief.

  • Microfiber material soft and absorbent
  • Feels significantly cooler than ambient air
  • Easily activated by running under water for one minute
  • ANSI certified 150D Oxford Polyester
  • ANSI-compliant tape with contrasting color

Specifications

Material Microfiber-lined Polyester
Product Type Ranger Hat
Color Lime
For Use With (Application) Road Construction, Utility Crew, Construction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.