Learn More
Electron Microscopy Sciences ChillDown Universal 250ml Beaker Cooler
SDP
Catalog No. 501902672
Encompass_Preferred
Rapidly cool agarose mixtures and other solutions with this versatile product. Chill molten agarose down to 60 degree C in minutes. Rapid cooling of agarose prevents an early-polymerized layer on top of the solution, which interferes with results. Fits any 250 mL beaker. Simply leave in freezer overnight and it will be ready for use.
Catalog No. 50-190-2672 Supplier Electron Microscopy Sciences Supplier No. 7751801
Rapidly cool agarose mixtures and other solutions with this versatile product. Chill molten agarose down to 60 degree C in minutes. Rapid cooling of agarose prevents an early-polymerized layer on top of the solution, which interferes with results. Fits any 250 mL beaker. Simply leave in freezer overnight and it will be ready for use.