Learn More
CHAMELEON COOLER CORP Chilled Blood Specimen Transporter
Catalog No. NC0522792
Encompass_Preferred
Chilled blood specimen transporter that remains between 32-34 degrees F for up to 6 hours after an overnight freeze. Holds up to six tubes: Clips onto the side of the phlebotomy tray: Color indicator signals at a glance whether the specimens are within a safe non metabolizing temperature: Blue when frozen and white at room temperature.
Catalog No. NC0522792 Supplier CHAMELEON COOLER CORP Supplier No. VNCC3255
Chilled blood specimen transporter that remains between 32-34 degrees F for up to 6 hours after an overnight freeze. Holds up to six tubes: Clips onto the side of the phlebotomy tray: Color indicator signals at a glance whether the specimens are within a safe non metabolizing temperature: Blue when frozen and white at room temperature.