Learn More
Polyscience DuraChill® 1/2HP Recirculating Chiller with Positive Displacement Pump (-10 to 70C); 240V, 50Hz
SDP
Catalog No. NC1814303
Encompass_Preferred
PolyScience™ DuraChill Recirculating Chiller 1/2HP Positive Displacement Pump -10 to 70C Working Temperature RangeTemperature Stability: 0.1CDiplayable Temperature Units: C or FDisplayable Pressure Units: PSI or kPaConnectivity Options: RS-232, USB Serial Port Emulation, USB TMC, Dry Contact On/Off/StatusOperating Languages: English, Spanish, French, German, MandarinCooling Capacity at 20C: 1840WPump Pressure Range: 20 to 83 psi / 138 to 572 kPa2.0 gpm / 7.6 lpm Maximum Flow Rate240V, 50HzFeatures: Large, full-color touchscreen display with intuitive interface and support for five different languages Compact, portable design takes up less floor space Cooling at ambient temperatures as high as 30C Whispercool noise reduction system DynamicFilter™ self-changing filter system Front fill reservoir Liquid level monitoring to prevent pump failure
Catalog No. NC1814303 Supplier Polyscience Supplier No. CA05A2P141AA1N
Add to Cart
Add to Cart
Request a Quote
PolyScience™ DuraChill Recirculating Chiller 1/2HP Positive Displacement Pump -10 to 70C Working Temperature RangeTemperature Stability: 0.1CDiplayable Temperature Units: C or FDisplayable Pressure Units: PSI or kPaConnectivity Options: RS-232, USB Serial Port Emulation, USB TMC, Dry Contact On/Off/StatusOperating Languages: English, Spanish, French, German, MandarinCooling Capacity at 20C: 1840WPump Pressure Range: 20 to 83 psi / 138 to 572 kPa2.0 gpm / 7.6 lpm Maximum Flow Rate240V, 50HzFeatures: Large, full-color touchscreen display with intuitive interface and support for five different languages Compact, portable design takes up less floor space Cooling at ambient temperatures as high as 30C Whispercool noise reduction system DynamicFilter™ self-changing filter system Front fill reservoir Liquid level monitoring to prevent pump failure