Learn More
Polyscience DuraChill® 1 HP Recirculating Chiller with Positive Displacement Pump (-10 to 70C); 240V, 50Hz
SDP
Supplier:  Polyscience CA10A2P241AA1N
Encompass_Preferred
PolyScience™ DuraChill Recirculating Chiller1HP Positive Displacement Pump-10 to 70C Working Temperature RangeTemperature Stability: 0.1CDiplayable Temperature Units: C or FDisplayable Pressure Units: PSI or kPaConnectivity Options: RS-232, USB Serial Port Emulation, USB TMC, Dry Contact On/Off/StatusOperating Languages: English, Spanish, French, German, MandarinCooling Capacity at 20C: 2650WPump Pressure Range: 20 to 83 psi / 138 to 572 kPa2.9 gpm (7.6 l/min) Maximum Flow Rate240V, 50HzFeatures: Large, full-color touchscreen display with intuitive interface and support for five different languages Compact, portable design takes up less floor space Cooling at ambient temperatures as high as 30C Whispercool noise reduction system DynamicFilter™ self-changing filter system Front fill reservoir User-adjustable temperature, pressure, and flow rate alarms
Catalog No. NC1814307
Only null left
Add to Cart
Request a Quote