Learn More
Polyscience DuraChill® 1HP Recirculating Chiller with Turbine Pump (-10 to 70C); 230V, 60Hz
SDP
Catalog No. NC1814308
Encompass_Preferred
PolyScience™ DuraChill Recirculating Chiller1HP Turbine Pump-10 to 70C Working Temperature RangeTemperature Stability: 0.1CDiplayable Temperature Units: C or FDisplayable Pressure Units: PSI or kPaConnectivity Options: RS-232, USB Serial Port Emulation, USB TMC, Dry Contact On/Off/StatusOperating Languages: English, Spanish, French, German, MandarinCooling Capacity at 20C: 2900WPump Pressure Range: 20 to 90 psi / 138 to 689 kPa3.5 gpm / 13.2 lpm Maximum Flow Rate230V, 60HzFeatures: Large, full-color touchscreen display with intuitive interface and support for five different languages Compact, portable design takes up less floor space Cooling at ambient temperatures as high as 30C Whispercool noise reduction system DynamicFilter™ self-changing filter system Front fill reservoir Liquid level monitoring to prevent pump failure Ultraviolet anti-biological light system User-adjustable temperature, pressure, and flow rate alarms
Catalog No. NC1814308 Supplier Polyscience Supplier No. CA10A3T141AA1N
Add to Cart
Add to Cart
Request a Quote
PolyScience™ DuraChill Recirculating Chiller1HP Turbine Pump-10 to 70C Working Temperature RangeTemperature Stability: 0.1CDiplayable Temperature Units: C or FDisplayable Pressure Units: PSI or kPaConnectivity Options: RS-232, USB Serial Port Emulation, USB TMC, Dry Contact On/Off/StatusOperating Languages: English, Spanish, French, German, MandarinCooling Capacity at 20C: 2900WPump Pressure Range: 20 to 90 psi / 138 to 689 kPa3.5 gpm / 13.2 lpm Maximum Flow Rate230V, 60HzFeatures: Large, full-color touchscreen display with intuitive interface and support for five different languages Compact, portable design takes up less floor space Cooling at ambient temperatures as high as 30C Whispercool noise reduction system DynamicFilter™ self-changing filter system Front fill reservoir Liquid level monitoring to prevent pump failure Ultraviolet anti-biological light system User-adjustable temperature, pressure, and flow rate alarms