Learn More
Polyscience DuraChill® 1/4HP Recirculating Chiller with Positive Displacement Pump (-10 to 70C); 120V, 60Hz
SDP
Catalog No. NC1814292
Encompass_Preferred
PolyScience™ DuraChill Recirculating Chiller1/4HP Positive Displacement Pump-10 to 70C Working Temperature RangeTemperature Stability: 0.1CDiplayable Temperature Units: C or FDisplayable Pressure Units: PSI or kPaConnectivity Options: RS-232, USB Serial Port Emulation, USB TMC, Dry Contact On/Off/StatusOperating Languages: English, Spanish, French, German, MandarinCooling Capacity at 20C: 700WPump Pressure Range: 45 to 100 psi / 310 to 689 kPa3.5 gpm / 13.2 lpm Maximum Flow Rate120V, 60HzFeatures: Large, full-color touchscreen display with intuitive interface and support for five different languages Compact, portable design takes up less floor space Cooling at ambient temperatures as high as 30C Whispercool noise reduction system DynamicFilter™ self-changing filter system Front fill reservoir
Catalog No. NC1814292 Supplier Polyscience Supplier No. CA02A1P141AA1N
Add to Cart
Add to Cart
Request a Quote
PolyScience™ DuraChill Recirculating Chiller1/4HP Positive Displacement Pump-10 to 70C Working Temperature RangeTemperature Stability: 0.1CDiplayable Temperature Units: C or FDisplayable Pressure Units: PSI or kPaConnectivity Options: RS-232, USB Serial Port Emulation, USB TMC, Dry Contact On/Off/StatusOperating Languages: English, Spanish, French, German, MandarinCooling Capacity at 20C: 700WPump Pressure Range: 45 to 100 psi / 310 to 689 kPa3.5 gpm / 13.2 lpm Maximum Flow Rate120V, 60HzFeatures: Large, full-color touchscreen display with intuitive interface and support for five different languages Compact, portable design takes up less floor space Cooling at ambient temperatures as high as 30C Whispercool noise reduction system DynamicFilter™ self-changing filter system Front fill reservoir