Learn More
THERMOTEK CHILLER-210 WATTS COOLING
SDP
Catalog No. NC1900950
Encompass_Preferred
Chiller - 210 watts cooling at 0°C delta T 21.75 psi (dead head) / 4 LPM (open flow) variable drive centrifugal Touch screen control Integrated fan speed control USB and RS232 communication Includes a set of mating male 3/8" ID fittings
Catalog No. NC1900950 Supplier THERMOTEK Supplier No. T257P20
Chiller - 210 watts cooling at 0°C delta T 21.75 psi (dead head) / 4 LPM (open flow) variable drive centrifugal Touch screen control Integrated fan speed control USB and RS232 communication Includes a set of mating male 3/8" ID fittings