Learn More
Polyscience Fitting, 1/2” male NPT x 3/8” (9.5 mm) Hose Barb, Brass, Straight
SDP
Catalog No. NC1722963
Encompass_Preferred
PolyScience™ PolyScience Fitting, 1/2” male NPT x 3/8” (9.5 mm) Hose Barb, Straight• 1/2” male NPT x 3/8” (9.5 mm) • BrassFor use with: • Circulating Baths• Refrigerated Circulating Baths• Heating Circulating Baths• Stainless Steel Open Bath Systems• Polycarbonate Open Bath Systems• Specialty Products• Calibration Bath• 75L Refrigerated Circulating Bath
Catalog No. NC1722963 Supplier Polyscience Supplier No. 776196
PolyScience™ PolyScience Fitting, 1/2” male NPT x 3/8” (9.5 mm) Hose Barb, Straight• 1/2” male NPT x 3/8” (9.5 mm) • BrassFor use with: • Circulating Baths• Refrigerated Circulating Baths• Heating Circulating Baths• Stainless Steel Open Bath Systems• Polycarbonate Open Bath Systems• Specialty Products• Calibration Bath• 75L Refrigerated Circulating Bath