Learn More
Cole-Parmer Cole-Parmer Polystat low-temperature immersion chiller; flex probe; 120V
Catalog No. NC1511271 Shop All Cole-Parmer Products
Encompass_Preferred
Quickly cool the contents of your bath with these immersion chillers. Digital display of bath temperature with 0.1°C resolution, and a 100Ω platinum RTD sensor for increased temperature stability. RTD temperature sensor includes a 6"L x 1/8" diameter stainless steel sheath and a 5-ft (1.5-m) cable. Probe can be inserted into your bath at ambient temperature—there’s no need to wait until your bath reaches subzero temperatures. A generous 60" of insulated tubing (11/2" OD) is between the probe and the cabinet.
Catalog No. NC1511271 Supplier Cole-Parmer Supplier No. 1457542
Add to Cart
Add to Cart
Request a Quote
Quickly cool the contents of your bath with these immersion chillers. Digital display of bath temperature with 0.1°C resolution, and a 100Ω platinum RTD sensor for increased temperature stability. RTD temperature sensor includes a 6"L x 1/8" diameter stainless steel sheath and a 5-ft (1.5-m) cable. Probe can be inserted into your bath at ambient temperature—there’s no need to wait until your bath reaches subzero temperatures. A generous 60" of insulated tubing (11/2" OD) is between the probe and the cabinet.