Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Catalog No. 13265245 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Plumbing Package
Tubing
Tubing Kit
For Use With (Equipment):
ThermoFlex 900 to 10000
ThermoTemp, Polar, VersaCool
Size:
1/2 in. NPT, 50 ft.
6 mm ID per meter, 0.5 meter long
6 mm ID per meter, 1.0 meter long
Material:
Material for High Temperature
Polybraid
Stainless-steel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment) Size Material
13-265-245 Tubing Kit ThermoFlex 900 to 10000 1/2 in. NPT, 50 ft. Polybraid
13-265-244 Plumbing Package ThermoFlex 900 to 10000 1/2 in. NPT, 50 ft. Material for High Temperature
13-100-780 Tubing ThermoTemp, Polar, VersaCool 6 mm ID per meter, 1.0 meter long Stainless-steel
13-100-779 Tubing ThermoTemp, Polar, VersaCool 6 mm ID per meter, 0.5 meter long Stainless-steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-265-245 Supplier Thermo Scientific™ Supplier No. 611000000100
Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
  • Include flexible tubing, tubing insulation, and hose clamps
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ThermoFlex 900 to 10000
Type Tubing Kit
Includes 50 ft. of 5/8 in. ID Polybraid hose comes with 1/2 in. hose barbs, insulation and clamps
Material Polybraid
Size 1/2 in. NPT, 50 ft.