Learn More
Grainger Chime Strobe White
Catalog No. 502767545
Encompass_Preferred
The Edwards Signaling EG1-CVM-CHIME-STROBE is a white 16" x 34.6" chime strobe. It has a low current draw and provides a minimum of 75 candela. The chime produces a minimum of 90 dBA at 10 feet. It is made of durable ABS plastic and has a weatherproof back box. The chime strobe is ETL listed and meets ADA signaling requirements. With an operating temperature range of 32°F to 120°F, this device is well-suited for indoor or outdoor signaling applications.
Catalog No. 50-276-7545 Supplier Grainger Supplier No. 16X346
The Edwards Signaling EG1-CVM-CHIME-STROBE is a white 16" x 34.6" chime strobe. It has a low current draw and provides a minimum of 75 candela. The chime produces a minimum of 90 dBA at 10 feet. It is made of durable ABS plastic and has a weatherproof back box. The chime strobe is ETL listed and meets ADA signaling requirements. With an operating temperature range of 32°F to 120°F, this device is well-suited for indoor or outdoor signaling applications.