Learn More
Grainger Chime White
Catalog No. 502766688
Encompass_Preferred
The Edwards Signaling 24V DC Chime is an electronic signaling device that produces an audible chime tone. It operates on 24V DC and draws 30mA. The chime provides a loud 80 decibel tone in a compact 6 x 2.75 x 1.25 inch ABS plastic housing. It can be flush mounted on walls or ceilings. The chime is commonly used for doorbells, nurse call stations, assembly lines, and other signaling applications. With an operating temperature range of -4 to 158°F, this versatile chime is suitable for indoor or outdoor use. The chime's dependable solid state circuitry and weatherproof construction make it a durable and reliable signaling solution.
Catalog No. 50-276-6688 Supplier Grainger Supplier No. 16X345
The Edwards Signaling 24V DC Chime is an electronic signaling device that produces an audible chime tone. It operates on 24V DC and draws 30mA. The chime provides a loud 80 decibel tone in a compact 6 x 2.75 x 1.25 inch ABS plastic housing. It can be flush mounted on walls or ceilings. The chime is commonly used for doorbells, nurse call stations, assembly lines, and other signaling applications. With an operating temperature range of -4 to 158°F, this versatile chime is suitable for indoor or outdoor use. The chime's dependable solid state circuitry and weatherproof construction make it a durable and reliable signaling solution.