Learn More
Creative Biomart Chimeric Chagas
Catalog No. 502084406
Encompass_Preferred
Recombinant Chimeric Chagas Multiantigen (MACH) (Trypanosoma cruzi) produced in E.Coli, is a polypeptide chain of 87 a.a. with epitopes PEP-2, TcD, TcE and SAPA. The protein is fused to a 6-His tag and having an Mw of 9.9kDa.
Catalog No. 50-208-4406 Supplier Creative Biomart Supplier No. CCS028
Recombinant Chimeric Chagas Multiantigen (MACH) (Trypanosoma cruzi) produced in E.Coli, is a polypeptide chain of 87 a.a. with epitopes PEP-2, TcD, TcE and SAPA. The protein is fused to a 6-His tag and having an Mw of 9.9kDa.