Learn More
REVMAB BIOSCIENCES USA INC His-Tag [RMH01], Chimeric Human Monoclonal Antibody
Supplier:  REVMAB BIOSCIENCES USA INC 54116100
Encompass_Preferred
Anti-His-Tag Chimeric Human Monoclonal Antibody, Clone RMH01 (100UG) ; WB, ELISA validated; Product info: www.revmab.com
Catalog No. NC1728988