Learn More
Andwin Scientific CHINA MARKERS BLUECHINA MARKE
SDP
Catalog No. NC9639400
Encompass_Preferred
The blue Sharpie Peel-Off China Marker can write on any porous or non-porous material. This can be utilized to mark surfaces like glass, ceramics, and plastics. It can withstand moisture and water without fading. This product does not need any sharpening. The string on the marker can be peeled down in order to freshen the tips.
Catalog No. NC9639400 Supplier Andwin Scientific Supplier No. 52036B
The blue Sharpie Peel-Off China Marker can write on any porous or non-porous material. This can be utilized to mark surfaces like glass, ceramics, and plastics. It can withstand moisture and water without fading. This product does not need any sharpening. The string on the marker can be peeled down in order to freshen the tips.