Learn More
Sino Biological Recombinant Chinese hamster Clusterin Protein (His Tag)
Supplier:  Sino Biological 5A7667A08C1
Encompass_Preferred
A DNA sequence encoding the chinese hamster CLU (G3HNJ3-1) (Glu22-Glu447) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6610