Learn More
American Research Products Inc Chinese Hamster OVary CHO Ho
Catalog No. 5000326406
Encompass
Chinese Hamster OVary (CHO) Host Cell Protein (HCP) Residue ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 3 1 ng/mL Sample cell lysates Method Sandwich
Catalog No. 50-003-26406 Supplier American Research Products Inc Supplier No. CSBEQ33262CHO
Chinese Hamster OVary (CHO) Host Cell Protein (HCP) Residue ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 3 1 ng/mL Sample cell lysates Method Sandwich