Learn More
American Research Products Inc Chinese Hamster OVary CHO Ho
Supplier:  American Research Products Inc CSBEQ33263CHO
Encompass
Chinese Hamster OVary (CHO) Host Cell Protein (HCP) Residue ELISA Kit from CUSABIO Detection Range 3 125 ng/mL-400 ng/mL Sensitivity 0 78 ng/mL Sample cell culture media purification intermediates final products and cell lysates Method Sandwich
Catalog No. 50-003-28880