Learn More
Sino Biological Anti-PLBL2 / PLBD2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 5A7664MM23100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Chinese hamster PLBL2/PLBD2 (Catalog# 5A7664-A08C; G3I6T1-1; Leu38-Asp585). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3978