Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diagenode Inc ChIP Cross-link Gold, 250x concentrated solution, 600 μL

Catalog No. NC0928534
Encompass

The Diagenode ChIP Cross-link Gold (C01019027) is a specialized reagent that enhances chromatin immunoprecipitation (ChIP) assays by stabilizing protein-protein interactions. This innovative product addresses the limitations of traditional formaldehyde fixation, particularly for proteins not directly bound to DNA. It facilitates a sequential fixation protocol that stabilizes protein complexes before interlinking protein-DNA, yielding a more comprehensive view of chromatin architecture.

  • Application in chromatin immunoprecipitation (ChIP)
  • Enhances protein-protein cross-linking
  • Targets higher-order and dynamic protein interactions
  • Shipped at -20°C
  • Store at -20°C; for long term, store at -80°C
  • Volume of 600 μL
  • Avoid multiple freeze-thaw cycles
  • 250x concentrated solution

Catalog No. NC0928534 Supplier Diagenode Inc Supplier No. C01019027
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Diagenode ChIP Cross-link Gold (C01019027) is a specialized reagent that enhances chromatin immunoprecipitation (ChIP) assays by stabilizing protein-protein interactions. This innovative product addresses the limitations of traditional formaldehyde fixation, particularly for proteins not directly bound to DNA. It facilitates a sequential fixation protocol that stabilizes protein complexes before interlinking protein-DNA, yielding a more comprehensive view of chromatin architecture.

  • Application in chromatin immunoprecipitation (ChIP)
  • Enhances protein-protein cross-linking
  • Targets higher-order and dynamic protein interactions
  • Shipped at -20°C
  • Store at -20°C; for long term, store at -80°C
  • Volume of 600 μL
  • Avoid multiple freeze-thaw cycles
  • 250x concentrated solution

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.