Learn More
Zymo Research Corporation ChIP DNA Clean & Concentrator™ (50 Preps) w/ Zymo-Spin™ IC Columns (Capped)
SDP
Catalog No. 50444363
Encompass_Preferred
The Chromatin Immunoprecipitation (ChIP) DNA Clean & Concentrator™ provides a hassle-free method for the rapid purification and concentration of high quality DNA from any step in a standard ChIP protocol.
Catalog No. 50-444-363 Supplier Zymo Research Corporation Supplier No. D5205
The Chromatin Immunoprecipitation (ChIP) DNA Clean & Concentrator™ provides a hassle-free method for the rapid purification and concentration of high quality DNA from any step in a standard ChIP protocol.