Learn More
Active Motif ChIP-IT Express
SDP
Catalog No. NC9584888
Encompass
ChIP-IT Kits provide all of the critical components needed for successful ChIP in a single kit. ChIP-IT comes complete with positive & negative control antibodies and PCR primers, DNA purification columns, optimized buffers and a comprehensive manual, all of which have been proven to work in ChIP. For your convenience, kits are available in a variety of formats, with your choice of sonication or our exclusive enzymatic shearing, and either with or without positive & negative controls. This enables you to use the shearing method that works best for you, and to either receive human-specific controls with the kit or to purchase ChIP-IT Control Kits separately to help analyze your results and verify that your antibodies work in the species you choose.
Catalog No. NC9584888 Supplier Active Motif Supplier No. 53008
ChIP-IT Kits provide all of the critical components needed for successful ChIP in a single kit. ChIP-IT comes complete with positive & negative control antibodies and PCR primers, DNA purification columns, optimized buffers and a comprehensive manual, all of which have been proven to work in ChIP. For your convenience, kits are available in a variety of formats, with your choice of sonication or our exclusive enzymatic shearing, and either with or without positive & negative controls. This enables you to use the shearing method that works best for you, and to either receive human-specific controls with the kit or to purchase ChIP-IT Control Kits separately to help analyze your results and verify that your antibodies work in the species you choose.