Learn More
Active Motif ChIP-IT qPCR Analysis Kit
SDP
Catalog No. NC1851556
Encompass
The ChIP-IT qPCR Analysis Kit was designed to simplify ChIP qPCR analysis and data interpretation. The kit includes standard curve DNA, a standard curve primer pair, positive and negative control qPCR primer sets for human and mouse samples and an analysis spreadsheet that automatically normalizes and graphs the data. The normalization calculations take into consideration the amount of chromatin that was used in the ChIP reaction, the resuspension volume of the ChIP DNA and the primer efficiencies to introduce a level of consistency in the data output that enables a reliable comparison of many samples across multiple experiments. Based on the consistency of the data output, Active Motif is able to make specific recommendations in regards to data interpretation so that customers can reliably assess the quality of their ChIP DNA.
Catalog No. NC1851556 Supplier Active Motif Supplier No. 53029
The ChIP-IT qPCR Analysis Kit was designed to simplify ChIP qPCR analysis and data interpretation. The kit includes standard curve DNA, a standard curve primer pair, positive and negative control qPCR primer sets for human and mouse samples and an analysis spreadsheet that automatically normalizes and graphs the data. The normalization calculations take into consideration the amount of chromatin that was used in the ChIP reaction, the resuspension volume of the ChIP DNA and the primer efficiencies to introduce a level of consistency in the data output that enables a reliable comparison of many samples across multiple experiments. Based on the consistency of the data output, Active Motif is able to make specific recommendations in regards to data interpretation so that customers can reliably assess the quality of their ChIP DNA.