Learn More
Abcam ChIP Kit Magnetic - One Step, 48 Tests
Catalog No. 502582821
Encompass_Preferred
Abcam's ChIP Kit Magnetic - One Step (ab156907) is a complete set of optimized buffers and reagents to carry out a successful chromatin immunoprecipitation for the selective enrichment of a chromatin fraction containing specific DNA sequences from isolated chromatin, in order to investigate the interaction between proteins and DNA in a cell.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2821 Supplier Abcam Supplier No. AB15690748TEST
Abcam's ChIP Kit Magnetic - One Step (ab156907) is a complete set of optimized buffers and reagents to carry out a successful chromatin immunoprecipitation for the selective enrichment of a chromatin fraction containing specific DNA sequences from isolated chromatin, in order to investigate the interaction between proteins and DNA in a cell.
The product is subject to the following: Abcam Restricted Use Statement