Learn More
Abcam ChIP Kit - One Step, 48 Tests
Catalog No. 502584817
Encompass_Preferred
Abcam's ChIP Kit - One Step (ab117138) enables the user to enrich a chromatin fraction containing specific DNA sequences from isolated chromatin to be able to investigate the interaction between proteins and DNA.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4817 Supplier Abcam Supplier No. AB11713848TEST
Abcam's ChIP Kit - One Step (ab117138) enables the user to enrich a chromatin fraction containing specific DNA sequences from isolated chromatin to be able to investigate the interaction between proteins and DNA.
The product is subject to the following: Abcam Restricted Use Statement