Learn More
Abcam ChIP Kit - One Step, 96 Tests
Catalog No. 502584816
Encompass_Preferred
Abcam's ChIP Kit - One Step (ab117138) enables the user to enrich a chromatin fraction containing specific DNA sequences from isolated chromatin to be able to investigate the interaction between proteins and DNA.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4816 Supplier Abcam Supplier No. AB11713896TEST
Abcam's ChIP Kit - One Step (ab117138) enables the user to enrich a chromatin fraction containing specific DNA sequences from isolated chromatin to be able to investigate the interaction between proteins and DNA.
The product is subject to the following: Abcam Restricted Use Statement