Learn More
Enzo Life Sciences AMPINEXT™ ChIP-Seq High-Sensitivity Kit (Illumina) (12Reactions)
SDP
Catalog No. 502013796
Encompass_Preferred
The AMPINEXT™ ChIP-Seq High-Sensitivity Kit (Illumina) is a complete set of reagents required for carrying out a successful ChIP-Seq starting from mammalian cells or tissues. The kit is designed to selectively enrich a chromatin fraction containing specific DNA sequences from various species, particularly mammals, and to prepare a ChIP-Seq library for next generation sequencing using Illumina platforms such as Illumina Genome Analyzer II, HiSeq and MiSeq systems. The optimized protocol and components of the kit allow capture of low abundance protein/DNA complexes with minimized non-specific background levels and the ability to construct both non-barcoded (singleplexed) and barcoded (multiplexed) ChIP-Seq libraries quickly with reduced bias. Assay Time: <7 hours.
Catalog No. 50-201-3796 Supplier Enzo Life Sciences Supplier No. ENZGEN5090012
The AMPINEXT™ ChIP-Seq High-Sensitivity Kit (Illumina) is a complete set of reagents required for carrying out a successful ChIP-Seq starting from mammalian cells or tissues. The kit is designed to selectively enrich a chromatin fraction containing specific DNA sequences from various species, particularly mammals, and to prepare a ChIP-Seq library for next generation sequencing using Illumina platforms such as Illumina Genome Analyzer II, HiSeq and MiSeq systems. The optimized protocol and components of the kit allow capture of low abundance protein/DNA complexes with minimized non-specific background levels and the ability to construct both non-barcoded (singleplexed) and barcoded (multiplexed) ChIP-Seq libraries quickly with reduced bias. Assay Time: <7 hours.