Learn More
Abcam CHIR-124, 50MG
Catalog No. 502671867
Encompass_Preferred
MW 419.9 Da, Purity >98%. Novel and potent Chk1 inhibitor (IC₅₀ = 0.3 nM), 2,000-fold more potent than Chk2 (IC₅₀ = 0.7 μM). Also, potently targets other kinases such as FLT3 (IC₅₀ = 5.8 nM), PDGFR (IC₅₀ = 6.6 nM), GSK-3 (IC₅₀ = 23.3 nM) and Fyn (IC₅₀ = 98.8 nM) .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1867 Supplier Abcam Supplier No. AB27361550MG
MW 419.9 Da, Purity >98%. Novel and potent Chk1 inhibitor (IC₅₀ = 0.3 nM), 2,000-fold more potent than Chk2 (IC₅₀ = 0.7 μM). Also, potently targets other kinases such as FLT3 (IC₅₀ = 5.8 nM), PDGFR (IC₅₀ = 6.6 nM), GSK-3 (IC₅₀ = 23.3 nM) and Fyn (IC₅₀ = 98.8 nM) .
The product is subject to the following: Abcam Restricted Use Statement