Learn More
Selleck Chemical LLC CHIR-98014 5mg 252935-94-7 CT98014
SDP
Supplier:  Selleck Chemical LLC S27455MG
Encompass_Preferred
CHIR-98014 (CT98014) is a potent GSK-3?/? inhibitor with IC50 of 0.65 nM/0.58 nM in cell-free assays, with the ability to distinguish GSK-3 from its closest homologs Cdc2 and ERK2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0595345