Learn More
Medchemexpress LLC HY-10182 10mg Medchemexpress, CHIR-99021 CAS:252917-06-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1018210MG
Encompass_Preferred
Medchemexpress, HY-10182 10mg CHIR-99021 CAS:252917-06-9 CHIR-99021 (CT99021) is a potent and selective GSK-3α/β inhibitor with IC50s of 10 nM and 6.7 nM. CHIR-99021 shows >500-fold selectivity for GSK-3 over CDC2, ERK2 and other protein kinases. CHIR-99021 is also a potent Wnt/β-catenin signaling pathway activator. CHIR-99021 enhances mouse and human embryonic stem cells self-renewal. CHIR-99021 induces autophagy[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8683
 Restricted Item