Learn More
BIOGEMS INTERNATIONAL INC CHIR 99021 10mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 252069110MG
Encompass
CHIR 99021 is an aminopyrimidine derivate that is a selective and potent inhibitor of glycogen synthase kinase 3 (GSK-3). It is used to maintain human and mouse stem cell lines and, in combination with other reagents, to chemically reprogram somatic cells into stem cells.
Catalog No. 50-152-9959