Learn More
Medchemexpress LLC HY-10182 100mg , CHIR-99021 CT99021 CAS:252917-06-9 Purity:98%
SDP
Catalog No. 501492664
Encompass_Preferred
Medchemexpress, HY-10182 100mg CHIR-99021 CT99021 CAS:252917-06-9 Formula:C22H18Cl2N8 IC50: 10 nM/6.7 nM (GSK-3α/β) Purity:98% CHIR-99021 is a GSK-3α/β inhibitor with IC 50 of 10 nM/6.7 nM; > 500-fold selectivity for GSK-3 versus its closest homologs CDC2 and ERK2, as well as other protein kinases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2664 Supplier Medchemexpress LLC Supplier No. HY10182100MG
Medchemexpress, HY-10182 100mg CHIR-99021 CT99021 CAS:252917-06-9 Formula:C22H18Cl2N8 IC50: 10 nM/6.7 nM (GSK-3α/β) Purity:98% CHIR-99021 is a GSK-3α/β inhibitor with IC 50 of 10 nM/6.7 nM; > 500-fold selectivity for GSK-3 versus its closest homologs CDC2 and ERK2, as well as other protein kinases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.