Learn More
Selleck Chemical LLC CHIR-99021 (CT99021) 2mg 252917-06-9
SDP
Supplier:  Selleck Chemical LLC S12632MG
Encompass_Preferred
CHIR-99021 (CT99021) is a GSK-3? and GSK-3? inhibitor with IC50 of 10 nM and 6.7 nM, respectively. CHIR99201 does not exhibit cross-reactivity against cyclin-dependent kinases (CDKs) and shows a 350-fold selectivity toward GSK-3? compared to CDKs. CHIR99021 functions as a Wnt/?-catenin activator and induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1039484