Learn More
Medchemexpress LLC HY-10182A 10mg Medchemexpress, CHIR-99021 (monohydrochloride) CAS:1797989-42-4 Purity:>98%
SDP
Catalog No. 501967878
Encompass_Preferred
Medchemexpress, HY-10182A 10mg CHIR-99021 (monoHCl) CAS:1797989-42-4 CHIR-99021 monohydrochloride (CT99021 monohydrochloride) is a GSK-3α/β inhibitor with IC50 of 10 nM/6.7 nM; > 500-fold selectivity for GSK-3 versus its closest homologs CDC2 and ERK2, as well as other protein kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7878 Supplier Medchemexpress LLC Supplier No. HY10182A10MG
Medchemexpress, HY-10182A 10mg CHIR-99021 (monoHCl) CAS:1797989-42-4 CHIR-99021 monohydrochloride (CT99021 monohydrochloride) is a GSK-3α/β inhibitor with IC50 of 10 nM/6.7 nM; > 500-fold selectivity for GSK-3 versus its closest homologs CDC2 and ERK2, as well as other protein kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.