Learn More
Cayman Chemical CHIR99021, 25 mg, GSK3 inhibitor, ≥95% purity, crystalline solid, -20°C storage, CAS 252917-06-9, molecular formula C₂₂H₁₈Cl₂N₈
SDP
Supplier:  Cayman Chemical 1312225
Encompass_Preferred
CHIR99021, 25 mg, GSK3 inhibitor, ≥95% purity, crystalline solid, -20°C storage, CAS 252917-06-9, molecular formula C₂₂H₁₈Cl₂N₈
Catalog No. NC2469120