Learn More
Reprocell CHIR99021 IN SOL 2MG-10MM
Catalog No. NC0391842
Encompass
The aminopyrimidine CHIR99021 is the most selective inhibitor of glycogen synthase kinase 3i� (GSK-3i�) reported to date1,2. Unlike other potent inhibitors of GSK-3, CHIR99201 does not exhibit cross-reactivity against cyclin-dependent kinases (CDKs) and shows a 350-fold selectivity toward GSK-3i� compared to CDKs3. along with the elimination of differentiation-inducing signaling from mitogen-activated protein kinases, using CHIR99021 to block the activity of GSK-3i� enables the self-renewal of embryonic stem cells4. Stemoleculea, c CHIR99021 in Solution is a ready to use 10 mM stock solution for stem cell culture.
Catalog No. NC0391842 Supplier Reprocell Supplier No. 04000402
The aminopyrimidine CHIR99021 is the most selective inhibitor of glycogen synthase kinase 3i� (GSK-3i�) reported to date1,2. Unlike other potent inhibitors of GSK-3, CHIR99201 does not exhibit cross-reactivity against cyclin-dependent kinases (CDKs) and shows a 350-fold selectivity toward GSK-3i� compared to CDKs3. along with the elimination of differentiation-inducing signaling from mitogen-activated protein kinases, using CHIR99021 to block the activity of GSK-3i� enables the self-renewal of embryonic stem cells4. Stemoleculea, c CHIR99021 in Solution is a ready to use 10 mM stock solution for stem cell culture.