Learn More
FUJIFILM BIOSCIENCES INC CHIR99021 MF
Catalog No. 5000450276
Encompass_Preferred
CHIR99021 is a potent and selective GSK-3a/b inhibitor used to promote stem cell maintenance and signal pathway modulation
Catalog No. 50-004-50276 Supplier FUJIFILM BIOSCIENCES INC Supplier No. 03825443
Add to Cart
Add to Cart
CHIR99021 is a potent and selective GSK-3a/b inhibitor used to promote stem cell maintenance and signal pathway modulation