Learn More
Chiral Technologies, Inc. CHIRACEL ODR COLUMN 250X4.6MM
Supplier:  Chiral Technologies, Inc. 14625
Encompass
CHIRALCEL OD-R10micrometer Particle Size 4.6 x 250mm Product Type: Analytical Adsorbent: 3 5-dimethylphenyl carbamate Applications: Reversed Phase type of CHIRALCEL OD or CHIRALCEL OD-H
Catalog No. NC9413266