Learn More
Chiral Technologies, Inc. CHIRAL PAK AD-H1 4.6X15MM 5U
Catalog No. NC9819623
Encompass
CHIRALPAK AD-H Particle Size (micro): 5 Internal Diameter (mm): 4.6 Column Length (mm):150 Product Type: Analytical Adsorbent: Amylose tris (3 5-dimethylphenyl carbamate) Applications: Use on compounds with aromatic amide carbamate ester gr
Catalog No. NC9819623 Supplier Chiral Technologies, Inc. Supplier No. 19324
CHIRALPAK AD-H Particle Size (micro): 5 Internal Diameter (mm): 4.6 Column Length (mm):150 Product Type: Analytical Adsorbent: Amylose tris (3 5-dimethylphenyl carbamate) Applications: Use on compounds with aromatic amide carbamate ester gr