Learn More
Strem, An Ascensus Company CAS# 871130-18-6. 100mg. (8aR)-18-Hydroxy-8,9-diphenyl-18-oxide-diphenanthro[4,3-d:3',4'-f][1,3,2]dioxaphosphepin, 98%, (99% ee).
SDP
Catalog No. 502065455
Encompass
CAS# 871130-18-6. 100mg. (8aR)-18-Hydroxy-8,9-diphenyl-18-oxide-diphenanthro[4,3-d:3',4'-f][1,3,2]dioxaphosphepin, 98%, (99% ee). . Molecular Weight: 600.6. Molecular Formula: C40H25O4P. Color/form: off white to light yellow pwdr. Strem# 15-8554. http://www.strem.com/catalog/v/15-8554/
Catalog No. 50-206-5455 Supplier Strem, An Ascensus Company Supplier No. 158554100MG
CAS# 871130-18-6. 100mg. (8aR)-18-Hydroxy-8,9-diphenyl-18-oxide-diphenanthro[4,3-d:3',4'-f][1,3,2]dioxaphosphepin, 98%, (99% ee). . Molecular Weight: 600.6. Molecular Formula: C40H25O4P. Color/form: off white to light yellow pwdr. Strem# 15-8554. http://www.strem.com/catalog/v/15-8554/